Skip to Content
Merck
All Photos(2)

Key Documents

697915

Sigma-Aldrich

4-Oxazolecarboxaldehyde

97%

Synonym(s):

Oxazole-4-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3NO2
CAS Number:
Molecular Weight:
97.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

57-61 °C

functional group

aldehyde

SMILES string

O=Cc1cocn1

InChI

1S/C4H3NO2/c6-1-4-2-7-3-5-4/h1-3H

InChI key

JBWIIXBEPINWPB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nadia Q Haj et al.
ACS omega, 5(23), 13948-13954 (2020-06-23)
Recently, chemical modifications of chitosan (CS) have attracted the attention of scientific researchers due to its wide range of applications. In this research, chitin (CH) was extracted from the scales of Cyprinus carpio fish and converted to CS by three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service