Skip to Content
Merck
All Photos(1)

Key Documents

687596

Sigma-Aldrich

(11bR)-(–)-4,4-Dibutyl-4,5-dihydro-2,6-bis(3,4,5-trifluorophenyl)-3H-dinaphth[2,1-c:1′,2′-e]azepinium bromide

Nagase purity

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H36BrF6N
CAS Number:
Molecular Weight:
748.64
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

223-228 °C

functional group

fluoro

SMILES string

CCCC[N+]1(CCCC)Cc2c(cc3ccccc3c2-c4c(C1)c(cc5ccccc45)-c6cc(F)c(F)c(F)c6)-c7cc(F)c(F)c(F)c7

InChI

1S/C42H36F6N.BrH/c1-3-5-15-49(16-6-4-2)23-33-31(27-19-35(43)41(47)36(44)20-27)17-25-11-7-9-13-29(25)39(33)40-30-14-10-8-12-26(30)18-32(34(40)24-49)28-21-37(45)42(48)38(46)22-28;/h7-14,17-22H,3-6,15-16,23-24H2,1-2H3;1H/q+1;/p-1

InChI key

LOMUZNOWQYZTRP-UHFFFAOYSA-M

Application

Potent phase transfer organocatalyst for asymmetric α-alkylation of glycine derivatives at extremely low loadings.

Features and Benefits

Maruoka phase-transfer catalysts are:
  • Operationally straightforward
  • Environmentally friendly
  • Relatively mild reaction conditions
  • Reactions conducted in aqueous media
  • Low catalyst loading

Legal Information

Product of Nagase & Co. Ltd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kitamura, M. et al.
Tetrahedron Asymmetry, 17, 603-603 (2006)
Organocatalysis
Aldrich Chemfiles, 7(9), 19-19 (2007)

Articles

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

Related Content

Designer chiral phase-transfer catalysts by Professor Maruoka advance various asymmetric transformations for diverse applications.

Designer chiral phase-transfer catalysts by Professor Maruoka advance various asymmetric transformations for diverse applications.

Designer chiral phase-transfer catalysts by Professor Maruoka advance various asymmetric transformations for diverse applications.

Designer chiral phase-transfer catalysts by Professor Maruoka advance various asymmetric transformations for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service