678732
Boron trichloride solution
1.0 M in toluene
Synonym(s):
Boron chloride, Trichloroborane
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
BCl3
CAS Number:
Molecular Weight:
117.17
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
concentration
1.0 M in toluene
density
0.909 g/mL at 25 °C
SMILES string
ClB(Cl)Cl
InChI
1S/BCl3/c2-1(3)4
InChI key
FAQYAMRNWDIXMY-UHFFFAOYSA-N
General description
Boron trichloride (BCl3) is a boron halide typically used as a reagent in organic synthesis for the cleavage of C-O bonds in ethers.
Application
Boron trichloride (BCl3) can be used as a reactant in the preperation of borazine-linked polymer [BLP-10(Cl)] by thermal decomposition with benzidine for gas storage and purification.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Corr. 1B - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-2.0 °F
Flash Point(C)
-18.9 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Highly selective CO 2/CH 4 gas uptake by a halogen-decorated borazine-linked polymer
Reich TE, et al.
Journal of Materials Chemistry, 22(27), 13524-13528 (2012)
Conversion of aromatic aldehydes to gem-dichlorides using boron trichloride. A new highly efficient method for preparing dichloroarylmethanes.
Kabalka G W and Wu Z
Tetrahedron Letters, 41(5), 579-581 (2000)
Juan Xie et al.
Carbohydrate research, 340(3), 481-487 (2005-02-01)
Boron trichloride has been found to promote selective deprotection of 1,2- or 1,3-cis oriented secondary benzyl ethers of per-benzylated C-glycosyl derivatives. The reactivity towards BCl(3) follows the order: C-4>or=C-2>C-6>C-3 for C-glucopyranosyl derivatives and C-3>or=C-4>C-6>C-2 for C-galactopyranosyl derivatives. Preparatively useful selective
Lei Zhang et al.
Journal of combinatorial chemistry, 7(4), 622-626 (2005-07-12)
A novel and efficient microwave-assisted, BCl3-mediated coupling reaction to synthesize salicylamide structures from phenols and isocyanates is described.
Jayaraman Selvakumar et al.
Organic & biomolecular chemistry, 8(18), 4056-4058 (2010-07-29)
Isoindoloisoquinalinone, pyrroloisoquinolinone and benzo[a]quinolizinone units are constructed via intramolecular cyclization of the methoxy substituted N-phenethylimides using BBr(3).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service