Skip to Content
Merck
All Photos(2)

Key Documents

656402

Sigma-Aldrich

2-Fluoroadenosine

97%

Synonym(s):

2-Fluoro-9-β-D-ribofuranosyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12FN5O4
CAS Number:
Molecular Weight:
285.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

240 °C (D) (lit.)

SMILES string

Nc1nc(F)nc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H12FN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1

InChI key

HBUBKKRHXORPQB-UUOKFMHZSA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergistic inhibition of platelet aggregation by forskolin plus PGE1 or 2-fluoroadenosine: effects of 2',5'-dideoxyadenosine and 5'-methylthioadenosine.
K C Agarwal et al.
Biochemical pharmacology, 31(22), 3713-3716 (1982-11-15)
Z H Chen et al.
Molecular pharmacology, 52(5), 903-911 (1997-11-14)
5'-Deoxy-5'-methylthioadenosine phosphorylase (MTAP) is involved in the salvage of adenine and methylthio moieties of 5'-deoxy-5'-methylthioadenosine, a byproduct of polyamine synthesis, to adenine nucleotides and methionine, respectively. The gene encoding MTAP, MTAP, is frequently codeleted along with the tumor suppressor gene
Mian M Alauddin et al.
Nuclear medicine and biology, 34(3), 267-272 (2007-03-27)
Many fluorinated analogues of adenosine nucleoside have been synthesized and studied as potential antitumor and antiviral agents. Earlier, we reported radiosynthesis of 2'-deoxy-2'-[(18)F]fluoro-1-beta-D-arabinofuranosyl-adenine ([(18)F]-FAA) and 3'-deoxy-3'-[(18)F]fluoro-1-beta-d-xylofuranosyl-adenine ([(18)F]FXA). Now, we report their in vivo studies including blood clearance, biodistribution and micro-PET
Luděk Eyer et al.
Antimicrobial agents and chemotherapy, 65(2) (2020-11-25)
Emerging flaviviruses are causative agents of severe and life-threatening diseases, against which no approved therapies are available. Among the nucleoside analogues, which represent a promising group of potentially therapeutic compounds, fluorine-substituted nucleosides are characterized by unique structural and functional properties.
U K Decking et al.
The American journal of physiology, 266(4 Pt 2), H1596-H1603 (1994-04-01)
Transport and phosphorylation of 2-fluoroadenosine (F-AR) were studied in human erythrocytes and porcine aortic endothelial cells by 19F-nuclear magnetic resonance (NMR) spectroscopy. F-AR (590 microM) added to a human erythrocyte suspension (15% hematocrit) was rapidly incorporated into adenine nucleotides at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service