Skip to Content
Merck
All Photos(1)

Key Documents

644153

Sigma-Aldrich

N-Methoxy-N-methyl-2,2,2-trifluoroacetamide

98%

Synonym(s):

N-Methyl-N-methoxytrifluoroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6F3NO2
CAS Number:
Molecular Weight:
157.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

53 °C/30 mmHg (lit.)

density

1.299 g/mL at 25 °C (lit.)

SMILES string

CON(C)C(=O)C(F)(F)F

InChI

1S/C4H6F3NO2/c1-8(10-2)3(9)4(5,6)7/h1-2H3

InChI key

JDBJZZTZYKUFFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.0 °F - closed cup

Flash Point(C)

47.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Courtney T Jollymore-Hughes et al.
Bioorganic & medicinal chemistry, 24(21), 5270-5279 (2016-09-18)
Association of cholinesterase with β-amyloid plaques and tau neurofibrillary tangles in Alzheimer's disease offers an opportunity to detect disease pathology during life. Achieving this requires development of radiolabelled cholinesterase ligands with high enzyme affinity. Various fluorinated acetophenone derivatives bind to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service