Skip to Content
Merck
All Photos(1)

Key Documents

64402

Sigma-Aldrich

Arquad® 2HT-75

Synonym(s):

Di(hydrogenated tallow)dimethylammonium chloride, Dimethyldioctadecylammonium chloride, mixture of homologs, Quaternium-18

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352000
NACRES:
NA.22

impurities

8-12% water

pH

6-9 (5% in isopropanol/water 1:1)

General description

Arquad® 2HT-75 or di(hydrogenated tallow)dimethylammonium chloride is a cationic surfactant used as an emulsifying, foaming, and wetting agent in various chemical industries. It is also used to form complexes with anionic species with exceptionally low water solubility results in flocculation, ion exchange, and decolorization characteristics that are useful in a variety of applications including water treatment, sugar refining, and the synthesis of organoclays.

Application

Arquad® 2HT-75 can be used as a dispersing agent in the preparation of nanocomposites to reduce surface tension between polymer and the filler. It is also used to develop bio-reactive organoclays because it is less toxic to soil microorganisms than other commonly used surfactants.

Legal Information

Arquad is a registered trademark of Akzo Nobel Chemicals B.V.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transfer batch blending, an innovative solvent/solid assisted method for melt compounding to achieve good dispersion quality for polymer-clay-nanocomposites
Hausner J, et al.
Composites Science and Technology, 114, 34-41 (2015)
Structural characterisation of Arquad 2HT-75 organobentonites: surface charge characteristics and environmental application
Sarkar B, et al.
Journal of Hazardous Materials, 195, 155-161 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service