Skip to Content
Merck
All Photos(1)

Key Documents

608963

Sigma-Aldrich

L-Lysine-2-15N hydrochloride

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(15NH2)CO2H·HCl
CAS Number:
Molecular Weight:
183.64
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]25/D +20.7°, c = 2 in 5 M HCl

technique(s)

protein expression: suitable

mp

263-264 °C (dec.) (lit.)

mass shift

M+1

SMILES string

Cl.NCCCC[C@H]([15NH2])C(O)=O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1/i8+1;

InChI key

BVHLGVCQOALMSV-WWQACITASA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service