Skip to Content
Merck
All Photos(1)

Key Documents

604429

Sigma-Aldrich

2-(1-Naphthyl)pentane-1,2-13C2

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C13H18
Molecular Weight:
200.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

mass shift

M+2

SMILES string

CCC[13CH]([13CH3])c1cccc2ccccc12

InChI

1S/C15H18/c1-3-7-12(2)14-11-6-9-13-8-4-5-10-15(13)14/h4-6,8-12H,3,7H2,1-2H3/i2+1,12+1

InChI key

MXJUSBINYALFBD-YZGYIGAXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service