Skip to Content
Merck
All Photos(1)

Key Documents

513954

Sigma-Aldrich

trans-Cinnamic-d7 acid

98 atom % D

Synonym(s):

3-Phenyl-d5-2-propenoic acid-2,3-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD=CDCO2H
CAS Number:
Molecular Weight:
155.20
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

bp

300 °C (lit.)

mp

132-135 °C (lit.)

mass shift

M+7

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])\C([2H])=C(/[2H])C(O)=O

InChI

1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+/i1D,2D,3D,4D,5D,6D,7D

InChI key

WBYWAXJHAXSJNI-UJMUNGNDSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Noelia Cambeiro-Pérez et al.
Frontiers in microbiology, 9, 2701-2701 (2018-12-14)
There are strong evidences that probiotics influence the immune status of the host, in a strain-specific manner, acting in the gastrointestinal tract. On the hypothesis that certain extracellular proteins and peptides from gut bacteria may mediate part of this immunomodulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service