Skip to Content
Merck
All Photos(1)

Key Documents

499234

Sigma-Aldrich

2-Acetyl-4-methylpyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO
CAS Number:
Molecular Weight:
135.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

mp

30-34 °C (lit.)

functional group

ketone

SMILES string

Cc1ccnc(c1)C(C)=O

InChI

1S/C8H9NO/c1-6-3-4-9-8(5-6)7(2)10/h3-5H,1-2H3

InChI key

HRVQMQWVGKYDCF-UHFFFAOYSA-N

General description

2-Acetyl-4-methylpyridine is a substituted pyridine that can be prepared from 2-bromo-4-methylpyridine.

Application

2-Acetyl-4-methylpyridine may be used in the synthesis of:
  • 3,5-bis(4,4"-dimethyl-2,2′:6′,2′′-terpyridin-4′-yl)-1-bromobenzene
  • 1,3-bis(4,4"-dimethyl-2,2′:6′,2′′-terpyridin-4′-yl)-5-methylbenzene
  • 2-(3′,3′-bis(ethylthio)-1′-oxoprop-2′-en-1′-yl)-4-methylpyridine
  • 2-(3′-(N,N-dimethylamino)-1′-oxoprop-2′-en-1′-yl)-4-methylpyridine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Construction of hexanuclear macrocycles by a coupling strategy from polyfunctionalized bis (terpyridines).
Eryazici I and Newkome GR
New. J. Chem., 33(2), 345-357 (2009)
Steric Control of Directional Isomerism in Dicopper (I) Helicates of Asymmetrically Substituted 2,2':6',2'':2'',6'''-Quaterpyridine Derivatives.
Constable EC, et al.
Journal of the American Chemical Society, 119(24), 5606-5617 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service