Skip to Content
Merck
All Photos(1)

Key Documents

47874

Sigma-Aldrich

Fmoc-β-Homolys(Boc)-OH

≥95.0% (HPLC)

Synonym(s):

(S)-7-(Boc-amino)-3-(Fmoc-amino)heptanoic acid, Nβ-Fmoc-Nω-Boc-L-β-homolysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O6
CAS Number:
Molecular Weight:
482.57
Beilstein:
8022293
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥95.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCCC[C@@H](CC(O)=O)NC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C27H34N2O6/c1-27(2,3)35-25(32)28-15-9-8-10-18(16-24(30)31)29-26(33)34-17-23-21-13-6-4-11-19(21)20-12-5-7-14-22(20)23/h4-7,11-14,18,23H,8-10,15-17H2,1-3H3,(H,28,32)(H,29,33)(H,30,31)/t18-/m0/s1

InChI key

SDBUQLGECIYUDT-SFHVURJKSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service