Skip to Content
Merck
All Photos(2)

Key Documents

473146

Sigma-Aldrich

Poly(dicyclopentadiene-co-p-cresol)

solid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

mol wt

degree of polymerization ~1

transition temp

Tg 92 °C

density

1.1 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(O)cc1.C2CC3C4CCC(C4)C3C2

InChI

1S/C10H12.C7H8O.C4H8/c1-2-9-7-4-5-8(6-7)10(9)3-1;1-6-2-4-7(8)5-3-6;1-4(2)3/h1-2,4-5,7-10H,3,6H2;2-5,8H,1H3;1H2,2-3H3

InChI key

UQQYHXZNSYZMEN-UHFFFAOYSA-N

General description

Poly(dicyclopentadiene-co-p-cresol) is a sterically hindered phenol.

Application

Poly(dicyclopentadiene-co-p-cresol) may be used as an antioxidant.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and preparation of high?performance thermoplastic vulcanizates based on blends of natural rubber and thermoplastic polyurethanes.
Kalkornsurapranee E, et al.
Journal of Applied Polymer Science, 128(4), 2358-2367 (2013)
Handbook for the Chemical Analysis of Plastic and Polymer Additives (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service