All Photos(1)
About This Item
Linear Formula:
HOC6H4C6H4CO2C2H5
CAS Number:
Molecular Weight:
242.27
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Recommended Products
Quality Level
Assay
98%
form
solid
mp
142-144 °C (dec.) (lit.)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
CCOC(=O)c1ccc(cc1)-c2ccc(O)cc2
InChI
1S/C15H14O3/c1-2-18-15(17)13-5-3-11(4-6-13)12-7-9-14(16)10-8-12/h3-10,16H,2H2,1H3
InChI key
FYXQIMAAEMCZLV-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service