Skip to Content
Merck
All Photos(2)

Key Documents

408573

Sigma-Aldrich

Diethyl (1-cyanoethyl)phosphonate

97%

Synonym(s):

(1-Cyanoethyl)phosphonic acid diethyl ester, NSC 135193

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CN)P(O)(OC2H5)2
CAS Number:
Molecular Weight:
191.16
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.432 (lit.)

bp

140-141 °C/11 mmHg (lit.)

density

1.085 g/mL at 25 °C (lit.)

functional group

nitrile
phosphonate

SMILES string

CCOP(=O)(OCC)C(C)C#N

InChI

1S/C7H14NO3P/c1-4-10-12(9,11-5-2)7(3)6-8/h7H,4-5H2,1-3H3

InChI key

UMLWEPGSWQNXQX-UHFFFAOYSA-N

Application

Reacant for:
  • Preparation of sphingosine phosphate receptor (S1P) agonists with N-cinnamyl-ß-alanine moiety
  • Preparation of benzyl- and benzoyldimethylaminopyridinones as HIV-1 reverse transcriptase inhibitors
  • Stereoselective preparation of cyano(trifluoromethyl)pentenenitriles via alkylation, followed by elimination reaction
  • Synthesis of [(methylenepiperidinyl) or (methylenepyrrolidinyl)fluorophenyl]oxazolidinone antibacterial agents
  • Horner-Wadsworth-Emmons olefination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service