Skip to Content
Merck
All Photos(2)

Key Documents

401420

Sigma-Aldrich

Calcium perchlorate tetrahydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(ClO4)2·4H2O
CAS Number:
Molecular Weight:
311.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
crystalline

Quality Level

Assay

99%

form

crystalline

reaction suitability

reagent type: oxidant

SMILES string

O.O.O.O.[Ca++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/Ca.2ClHO4.4H2O/c;2*2-1(3,4)5;;;;/h;2*(H,2,3,4,5);4*1H2/q+2;;;;;;/p-2

InChI key

YGFQNBSXMKXDTH-UHFFFAOYSA-L

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pawel Peter Bawol et al.
ChemSusChem, 14(1), 428-440 (2020-09-01)
In this study the fundamental understanding of the underlying reactions of a possible Ca-O2 battery using a DMSO-based electrolyte was strengthened. Employing the rotating ring disc electrode, a transition from a mixed process of O2 - and O2 2- formation
Yuanyuan He et al.
Astrobiology, 21(3), 279-297 (2020-12-12)
The Mars Organic Molecule Analyzer (MOMA) and Sample Analysis at Mars (SAM) instruments onboard the Exomars 2022 and Mars Science Laboratory rovers, respectively, are capable of organic matter detection and differentiating potentially biogenic from abiotic organics in martian samples. To
Xinyu Tian et al.
ChemPlusChem, 85(8), 1639-1645 (2020-08-05)
We report the synthesis, characterization, and photophysical and ion-binding properties of deep-red to near-infrared (NIR) fluorescent rhodamine derivatives, bearing two spirolactone rings and substitution of the oxygen atoms in the xanthene ring with sulfur atoms (1-S). The diastereoisomeric cis- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service