Skip to Content
Merck
All Photos(2)

Key Documents

381543

Sigma-Aldrich

N-Cyclohexylmaleimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

89-91 °C (lit.)

functional group

imide
maleimide

SMILES string

O=C1C=CC(=O)N1C2CCCCC2

InChI

1S/C10H13NO2/c12-9-6-7-10(13)11(9)8-4-2-1-3-5-8/h6-8H,1-5H2

InChI key

BQTPKSBXMONSJI-UHFFFAOYSA-N

General description

N-Cyclohexylmaleimide (CHMI, NCMI) is a cyclic imide. Diels-Alder reactions of 9-hydroxymethylanthracene with CHMI catalyzed by hydrophobic nanospace confined within the self-assembled Pd6 open cage bearing triimidazole walls have been reported. It undergoes Diels-Alder reactions with various aromatic hydrocarbons promoted by self-assembled coordination cage. This cage acts as nanometer-sized molecular flask and hence promotes this reaction. CHMI is reported to undergo radical polymerization readily under various polymerization conditions to afford poly(CHMI), having excellent thermal stability.

Application

N-Cyclohexylmaleimide (CHMI, NCMI) may be used for the preparation of hyperbranched copolymers of p-(chloromethyl)styrene (CMS) and NCMI, via atom transfer radical copolymerization reaction.
N-Cyclohexylmaleimide contaminated with N-cyclohexylmaleamic acid (less than 0.9 wt%) may be used for the heat-resistant poly(methyl methacrylate) resin free from coloration and for methyl methacrylate homopolymer. It may be used as monomer in the preparation of styrene/N-cyclohexylmaleimide copolymers with small polydispersities and controlled molecular weights, via free radical copolymerization. It may be used in the synthesis of hyperbranched copolymers by the atom transfer radical copolymerization with p-(chloromethyl)styrene catalyzed by CuCl/2,2′-bipyridine in cyclohexanone or anisole.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of N-cyclohexylmaleimide for heat-resistant transparent methacrylic resin.
Kita Y, et al.
Journal of Applied Polymer Science, 63(3), 363-368 (1997)
Increase in thermal stability of vinyl polymers through radical copolymerization with N-cyclohexylmaleimide.
Otsu T, et al.
Polymer International, 25(3), 179-184 (1991)
Living free radical donor-acceptor copolymerization of styrene and N-cyclohexylmaleimide and the synthesis of poly [styrene-co-(N-cyclohexylmaleimide)]/polystyrene block copolymers.
Schmidt-Naake G and Butz S.
Macromolecular Rapid Communications, 17(9), 661-665 (1996)
Bingchuan Wei et al.
Journal of chromatography. A, 1526, 104-111 (2017-10-29)
Reversed-phase liquid chromatography (RPLC) has been commonly used in IgG2 disulfide isoforms analysis. Recently, the columns packed with large pore superficially porous particles (SPP) have become available commercially. This work explores the application of this SPP technology in IgG2 disulfide
Hyperbranched copolymers of p-(chloromethyl) styrene and N-cyclohexylmaleimide synthesized by atom transfer radical polymerization.
Jiang X, et al.
Journal of Applied Polymer Science, 78(11), 1992-1997 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service