Skip to Content
Merck
All Photos(1)

Key Documents

333603

Sigma-Aldrich

6-Phenylhexanoic acid

98%

Synonym(s):

Benzenehexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)5CO2H
CAS Number:
Molecular Weight:
192.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.51 (lit.)

bp

201-202 °C/24 mmHg (lit.)

density

1.022 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCCCCc1ccccc1

InChI

1S/C12H16O2/c13-12(14)10-6-2-5-9-11-7-3-1-4-8-11/h1,3-4,7-8H,2,5-6,9-10H2,(H,13,14)

InChI key

JTXZPQIXIXYMDY-UHFFFAOYSA-N

General description

6-Phenylhexanoic acid is an arylalkanoic acid.

Application

6-Phenylhexanoic acid was employed as a model compound to investigate the effects of chromophore orientation and molecular conformation on surface-enhanced Raman scattering based on metal nanostructures. It was also employed as a substrate to study the stoichiometry of side chain metabolism or complete mineralization of surrogate napthenic acids under methanogenic conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Leo Seballos et al.
The Journal of chemical physics, 125(23), 234706-234706 (2006-12-28)
Experimental studies have been carried out to gain a better understanding of the effects of chromophore orientation and molecular conformation on surface-enhanced Raman scattering (SERS) based on metal nanostructures. A series of alkanoic acids that contain a phenyl ring separated
F M Holowenko et al.
Water research, 35(11), 2595-2606 (2001-07-18)
Naphthenic acids (NAs) are a complex mixture of naturally occurring acyclic and cyclic aliphatic carboxylic acids in petroleum. In the Athabasca oil sands. NAs have been identified as the largest component of dissolved organic matter in the tailings waters from
Keishi Yamasaki et al.
Journal of pharmaceutical sciences, 106(9), 2860-2867 (2017-05-01)
Sodium 4-phenylbutyrate (PB) is clinically used as a drug for treating urea cycle disorders. Recent research has shown that PB also has other pharmacologic activities, suggesting that it has the potential for use as a drug for treating other disorders.
Kamal Adhikari et al.
Biomolecules, 10(5) (2020-05-14)
Plant type III polyketide synthases produce diverse bioactive molecules with a great medicinal significance to human diseases. Here, we demonstrated versatility of a stilbene synthase (STS) from Pinus Sylvestris, which can accept various non-physiological substrates to form unnatural polyketide products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service