Skip to Content
Merck
All Photos(3)

Documents

304166

Sigma-Aldrich

2-Bromo-1,3,2-benzodioxaborole

97%

Synonym(s):

B-Bromocatechol-borane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BBrO2
CAS Number:
Molecular Weight:
198.81
Beilstein:
2445073
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

76 °C/9 mmHg (lit.)

mp

51-53 °C (lit.)

storage temp.

2-8°C

SMILES string

Brb1oc2ccccc2o1

InChI

1S/C6H4BBrO2/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H

InChI key

SAILTPCYIYNOEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service