Skip to Content
Merck
All Photos(2)

Key Documents

302341

Sigma-Aldrich

Acetoxyacetic acid

99%

Synonym(s):

α-Acetoxyacetic acid, (Acetyloxy)acetic acid, 2-(Acetyloxy)acetic acid, 2-Hydroxyacetic acid acetate, Acetylglycolic acid, O-Acetylglycolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CO2H
CAS Number:
Molecular Weight:
118.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

bp

141-142 °C/12 mmHg (lit.)

mp

67-69 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

CC(=O)OCC(O)=O

InChI

1S/C4H6O4/c1-3(5)8-2-4(6)7/h2H2,1H3,(H,6,7)

InChI key

MLXDUYUQINCFFV-UHFFFAOYSA-N

General description

Acetoxyacetic acid is kinetically stable degradation product of oxidative degradation of 1,3-dialkylimidazolium ionic liquids in hydrogen peroxide/aceticacid aqueous medium assisted by ultrasonic chemical irradiation.[1] It is an intermediate formed during reactions of manganic acetate in glacial acetic acid with benzene (100°C), chlorobenzene, or toluene.[2]

Application

Acetoxyacetic acid was used in preparation of tert-butyl glycolate.[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuehui Li et al.
Dalton transactions (Cambridge, England : 2003), (19)(19), 1875-1880 (2007-08-19)
A highly efficient process for oxidative degradation of 1,3-dialkylimidazolium ionic liquids in hydrogen peroxide/acetic acid aqueous medium assisted by ultrasonic chemical irradiation is, for the first time, described. It is shown that more than 93% of the 1,3-dialkylimidazolium cation with
Intermediate steps in autoxidation: III. Reactions of MnIII acetate in acetic acid with benzene, chlorobenzene, and toluene.
Van der PRE, et al.
J. Catal., 10(1), 52-59 (1968)
Synthetic studies of pyridomycin. II. Synthesis of a model twelve-membered ring compound related to pyridomycin.
Kinoshita M and Awamura M.
Bulletin of the Chemical Society of Japan, 51(3), 869-871 (1978)
Amberlyn M Wands et al.
ACS infectious diseases, 4(5), 758-770 (2018-02-08)
Cholera toxin (CT) enters host intestinal epithelia cells, and its retrograde transport to the cytosol results in the massive loss of fluids and electrolytes associated with severe dehydration. To initiate this intoxication process, the B subunit of CT (CTB) first

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service