Skip to Content
Merck
All Photos(2)

Key Documents

281964

Sigma-Aldrich

Butyl butyrate

98%

Synonym(s):

1-Butyl butyrate, Butyl butanoate, Butyric acid n-butyl ester, n-Butyl butanoate, n-Butyl butyrate, n-Butyl n-butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COO(CH2)3CH3
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747101
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.406 (lit.)

bp

164-165 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCOC(=O)CCC

InChI

1S/C8H16O2/c1-3-5-7-10-8(9)6-4-2/h3-7H2,1-2H3

InChI key

XUPYJHCZDLZNFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl butyrate is an ester present in pineapple flavor and is very important for the food and beverages industries. Kinetcis of butyl butyrate synthesis by enzymatic esterification and trans-esterification in supercritical carbon dioxide with a lipase, Novozym 435 has been reported.

Application

Butyl butyrate can be used as a reactant to synthesize:
  • N



  • -(Phenylmethyl)butanamide by reacting with benzylamine via ester-amide exchange reaction in the presence of supported graphene oxide catalyst.
  • Cyclohexyl butyrate by acylation reaction with cyclohexanol using a ruthenium pincer PNN complex catalyst.
  • Butyl ether by triiron dodecacarbonyl catalyzed hydrosilylation reaction.

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

124.9 °F - closed cup

Flash Point(C)

51.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of synthesis of butyl butyrate by esterification and transesterification in supercritical carbon dioxide.
Varma MN and Madras G.
Journal of Chemical Technology and Biotechnology, 83(8), 1135-1144 (2008)
Unprecedented Ester-Amide Exchange Reaction Using Highly Versatile Two-Dimensional Graphene Oxide Supported Base Metal Nanocatalyst
Sharma RK, et al.
Industrial & Engineering Chemistry Research, 57(10), 3617-3627 (2018)
Ruthenium Pincer-Catalyzed Acylation of Alcohols Using Esters with Liberation of Hydrogen under Neutral Conditions
Gnanaprakasam B, et al.
advanced synthesis and catalysis, 352(18), 3169-3173 (2010)
Ruthenium Pincer-Catalyzed Acylation of Alcohols Using Esters with Liberation of Hydrogen under Neutral Conditions
Gnanaprakasam B, et al.
Advanced Synthesis & Catalysis, 352(18), 3169-3173 (2010)
Andréa B Martins et al.
Biotechnology progress, 29(6), 1416-1421 (2013-08-16)
Butyl butyrate is an ester present in pineapple flavor, which is very important for the food and beverages industries. In this work, the optimization of the reaction of butyl butyrate synthesis catalyzed by the immobilized lipase Lipozyme TL-IM was performed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service