Skip to Content
Merck
All Photos(1)

Documents

262056

Sigma-Aldrich

Dibromoborane dimethyl sulfide complex solution

1.0 M in methylene chloride

Synonym(s):

Dibromo(dimethyl sulfide)(hydro)boron solution, Dibromo(dimethyl sulfide)borane solution, Dibromoborane compound with dimethylsulfane solution, Dibromoborane dimethyl sulfide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BHBr2
CAS Number:
Molecular Weight:
233.76
Beilstein:
4449985
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in methylene chloride

density

1.415 g/mL at 25 °C

SMILES string

CSC.BrBBr

InChI

1S/C2H6S.BBr2H/c1-3-2;2-1-3/h1-2H3;1H

InChI key

PGSRDLGPKTVELT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Reductive elimination C-C bond forming reactions
  • Lithiation-borylation reaction with Hoppe′s lithiated carbamates
  • Preparation of boron analogues of uracil
  • Boronation reactions for formation of cyclic boronium salts
  • Hydroboration reactions
  • Ring opening of terminal epoxides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service