Skip to Content
Merck
All Photos(1)

Key Documents

260088

Sigma-Aldrich

3-Bromophenylacetonitrile

98%

Synonym(s):

3-Bromobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2CN
CAS Number:
Molecular Weight:
196.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

145-147 °C/10 mmHg (lit.)

mp

27-28 °C (lit.)

SMILES string

Brc1cccc(CC#N)c1

InChI

1S/C8H6BrN/c9-8-3-1-2-7(6-8)4-5-10/h1-3,6H,4H2

InChI key

UUZYFBXKWIQKTF-UHFFFAOYSA-N

Application

3-Bromophenylacetonitrile has been used in the synthesis of:
  • a series of aminoethylbiphenyls, novel 5-HT7 receptor ligands
  • 2-(1-cyano-1-(3-bromophenyl))methylidene-3-phenylthiazolidine-4,5-dione

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magalie Paillet-Loilier et al.
Bioorganic & medicinal chemistry letters, 17(11), 3018-3022 (2007-04-10)
The synthesis of a series of aminoethylbiphenyls as novel 5-HT(7) receptor ligands is described. The novel derivatives exhibit high affinity for the 5-HT(7) receptor with selectivity toward 5-HT(1A) receptor.
Uwe Albrecht et al.
Bioorganic & medicinal chemistry, 13(14), 4402-4407 (2005-05-21)
2-Alkylidenethiazolidine-4,5-diones were prepared by novel one-pot cyclizations of arylacetonitriles with isothiocyanates and ethyl 2-chloro-2-oxoacetate. The products show antibiotic activity against the Gram-positive bacteria Bacillus subtilis and Staphylococcus aureus.
Xin-Yang Zhang et al.
Journal of hazardous materials, 403, 124103-124103 (2020-12-04)
Ozonation might increase the risk of wastewater due to byproduct formation, especially in the presence of bromide. In this study, a new analytical method was developed to identify new brominated disinfection byproducts (Br-DBPs) during ozonation, using comprehensive two-dimensional gas chromatography-single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service