247146
2-Chlorothiophenol
99%
Synonym(s):
2-Chlorobenzenethiol
About This Item
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.6029 (lit.)
bp
205-206 °C/260 mmHg (lit.)
density
1.275 g/mL at 25 °C (lit.)
functional group
chloro
SMILES string
Sc1ccccc1Cl
InChI
1S/C6H5ClS/c7-5-3-1-2-4-6(5)8/h1-4,8H
InChI key
PWOBDMNCYMQTCE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
190.4 °F - closed cup
Flash Point(C)
88 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service