Skip to Content
Merck
All Photos(1)

Key Documents

238864

Sigma-Aldrich

(S)-(+)-1-Amino-2-propanol

97%

Synonym(s):

(+)-Isopropanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2NH2
CAS Number:
Molecular Weight:
75.11
Beilstein:
1718868
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.6 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

97%

optical activity

[α]20/D +18°, c = 1.8 in H2O

refractive index

n20/D 1.4437 (lit.)

bp

160 °C (lit.)

mp

24-26 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

C[C@H](O)CN

InChI

1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3/t3-/m0/s1

InChI key

HXKKHQJGJAFBHI-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(+)-1-Amino-2-propanol is a chiral amino alcohol.

Application

(S)-(+)-1-Amino-2-propanol may be used in the preparation of S-(+)-1-(2,3-naphthalimido)-2-propanol.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Enantioselective asymmetric hydrogenation of a-phthalimide ketone: an efficient entry to enantiomerically pure amino alcohols.
Lei A, et al.
Journal of the American Chemical Society, 126(6), 1626-1627 (2004)
S-(+)-1-Methyl-2-(2, 3-naphthalimido) ethyl Trifluoromethanesulfonate as a Fluorescence Chiral Labeling Reagent for Carboxylic Acid Enantiomers.
Yasaka Y, et al.
Analytical Sciences, 11(2), 295-297 (1995)
D Catone et al.
The Journal of chemical physics, 127(14), 144312-144312 (2007-10-16)
Valence band and C 1s core-level photoelectron spectra of S-(+)-2-amino-1-propanol (alaninol) and S-(+)-1-amino-2-propanol (isopropanolamine) have been studied by means of synchrotron radiation photoelectron spectroscopy in gas phase. The alaninol, the reduced derivative of the alanine, is a good test system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service