Skip to Content
Merck
All Photos(1)

Key Documents

222143

Sigma-Aldrich

4-Pentylbenzoyl chloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C6H4COCl
CAS Number:
Molecular Weight:
210.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.53 (lit.)

density

1.036 g/mL at 25 °C (lit.)

SMILES string

CCCCCc1ccc(cc1)C(Cl)=O

InChI

1S/C12H15ClO/c1-2-3-4-5-10-6-8-11(9-7-10)12(13)14/h6-9H,2-5H2,1H3

InChI key

FBBRKYLXMNQFQU-UHFFFAOYSA-N

Application

4-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rama Mukherjee et al.
Bioorganic & medicinal chemistry letters, 14(12), 3169-3172 (2004-05-20)
New 3-O-acyl, 3-benzylidene, 3-hydrazone, 3-hydrazine, 17-carboxyacryloyl ester derivatives of betulinic acid (2-6, 8-11, 13, 17, 18, 21, and 22) were synthesized and evaluated in vitro for anti-angiogenic activity on endothelial cell cytotoxicity, specificity, and tube-formation ability. All derivatives reported here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service