Skip to Content
Merck
All Photos(4)

Key Documents

204439

Sigma-Aldrich

Sodium chloride

99.999% trace metals basis

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
solid
solubility:
water: soluble (358 g/l at 20 °C (68 °F))

vapor pressure

1 mmHg ( 865 °C)

Quality Level

Assay

99.999% trace metals basis

form

solid

impurities

≤15.0 ppm Trace Metal Analysis

pH

7

mp

801 °C (lit.)

solubility

water: soluble (358 g/l at 20 °C (68 °F))

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium chloride is a white crystalline solid highly soluble in water. High-purity sodium chloride is commonly used in the pharmaceutical and semiconductor industry.

Application

Facilitates the cross-coupling of organostannanes with iodides without using palladium.
Sodium chloride can be used:
  • As an aqueous electrolyte in batteries.
  • To detect trace elements using inductively coupled plasma mass spectrometry.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new procedure for the determination of 21 macro- and trace elements in human fetal urine using an inductively coupled plasma mass spectrometry with dynamic reaction cell (ICP-DRC-MS) equipped with a micro-flow nebulizer
Adam Sajnog, et al.
Talanta, 222, 121672-121672 (2021)
Developments in electrode materials and electrolytes for aluminium?air batteries
D.R. Egan, et al.
Journal of Power Sources, 236, 293-310 (2013)
Copper- and Manganese-Catalyzed Cross-Coupling of Organostannanes with Organic Iodides in the Presence of Sodium Chloride.
Suk-Ku Kang et al.
The Journal of organic chemistry, 62(13), 4208-4209 (1997-06-27)
Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
Alexander W Wyatt et al.
Genome biology, 15(8), 426-426 (2014-08-27)
Genomic analyses of hundreds of prostate tumors have defined a diverse landscape of mutations and genome rearrangements, but the transcriptomic effect of this complexity is less well understood, particularly at the individual tumor level. We selected a cohort of 25

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service