Skip to Content
Merck
All Photos(1)

Key Documents

196819

Sigma-Aldrich

Tetrafluorophthalonitrile

95%

Synonym(s):

1,2-Dicyano-3,4,5,6-tetrafluorobenzene, 3,4,5,6-Tetrafluoro-1,2-benzenedicarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F4(CN)2
CAS Number:
Molecular Weight:
200.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

81-86 °C (lit.)

SMILES string

Fc1c(F)c(F)c(C#N)c(C#N)c1F

InChI

1S/C8F4N2/c9-5-3(1-13)4(2-14)6(10)8(12)7(5)11

InChI key

OFLRJMBSWDXSPG-UHFFFAOYSA-N

General description

Tetrafluorophthalonitrile reacts with:
  • copper, copper (I) chloride or copper (II) chloride to yield copper (II) hexadecafluorophthalocyanine
  • potassium salt of 2-hydroxyhexafluoro-2-propylbenzene to yield 2-phenyl-2-(3,4-dicyano- trifluorophenoxy) hexafluoropropane
  • dipotassium salt of 1,3-bis(2-hdroxyhexafluoro-2- propyl) benzene to yield fluorinated phthalonitrile resins

Application

Tetrafluorophthalonitrile was used in the synthesis of dichloro-subphthalocyanine dimers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of highly fluorinated phthalonitrile resins and cure studies.
Keller TM and Griffith JR.
Journal of Fluorine Chemistry, 13(4), 315-324 (1979)
Polyfluoroarenes. Part XI. Reactions of tetrafluorophthalonitrile with nucleophilic reagents.
Birchall JM, et al.
J. Chem. Soc. Sect. C, 3, 456-462 (1970)
Synthesis, separation, and characterization of the topoisomers of fused bicyclic subphthalocyanine dimers.
Christian G Claessens et al.
Angewandte Chemie (International ed. in English), 41(14), 2561-2565 (2002-08-31)
Thi Hai Quyen Nguyen et al.
Physical chemistry chemical physics : PCCP, 22(15), 7699-7709 (2020-03-07)
The transport of both electrons and ions in organic mixed ionic and electronic conductors such as phthalocyanines, is essential to allow redox reactions of entire films and, hence, to impart electrochromism. Thin films of a new type, tetrakis-perfluoroisopropyl-perfluoro phthalocyanine, F40PcCu
Dongkun Yu et al.
The journal of physical chemistry. B, 123(23), 4958-4966 (2019-05-24)
The concept of eutectic molecular liquids (EMLs) was defined, and a strategy to form EMLs based on noncovalent interactions was proposed. We verified the formation and investigated the properties, interaction sites, and interaction energies of the obtained 16 EMLs. Moreover

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service