184454
Boric acid gel
0.1-0.4 mm particle size
Synonym(s):
3-Aminophenylboronic acid, polymer-bound
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H12BNO3
CAS Number:
Molecular Weight:
205.02
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
reactivity: alcohol reactive
particle size
0.1-0.4 mm
density
0.6 g/mL at 25 °C (lit.)
SMILES string
CC(=C)C(=O)Nc1cccc(c1)B(O)O
InChI
1S/C10H12BNO3/c1-7(2)10(13)12-9-5-3-4-8(6-9)11(14)15/h3-6,14-15H,1H2,2H3,(H,12,13)
InChI key
GBBUBIKYAQLESK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Useful to capture avidin for boron-neutron capture therapy applications
Features and Benefits
Cross-linked polymer for the column chromatographic separation of mixtures whose components form complexes of varying stability with boric acid.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service