Skip to Content
Merck
All Photos(1)

Key Documents

163627

Sigma-Aldrich

Tetraethyl orthocarbonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(OC2H5)4
CAS Number:
Molecular Weight:
192.25
Beilstein:
1747401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.392 (lit.)

bp

159 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCOC(OCC)(OCC)OCC

InChI

1S/C9H20O4/c1-5-10-9(11-6-2,12-7-3)13-8-4/h5-8H2,1-4H3

InChI key

CWLNAJYDRSIKJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetraethyl orthocarbonate was used in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane. It was used in the synthesis of crosslinked poly(orthocarbonate)s used as organic solvent absorbent.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of crosslinked poly (orthosilicate) s as organic solvent absorbent.
Sonmez HB, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 49, 846-846 (2008)
Radical polymerization of unsaturated spiroorthocarbonate.
Tagoshi H and Endo T.
Journal of Polymer Science: Part A, General Papers, 27(4), 1415-1418 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service