Skip to Content
Merck
All Photos(2)

Key Documents

163619

Sigma-Aldrich

4-Benzyloxy-3-methoxybenzaldehyde

98%

Synonym(s):

O-Benzylvanillin, Vanillin benzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
242.27
Beilstein:
1464258
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

62-64 °C (lit.)

functional group

aldehyde
phenyl

SMILES string

[H]C(=O)c1ccc(OCc2ccccc2)c(OC)c1

InChI

1S/C15H14O3/c1-17-15-9-13(10-16)7-8-14(15)18-11-12-5-3-2-4-6-12/h2-10H,11H2,1H3

InChI key

JSHLOPGSDZTEGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N′-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

Application

4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of the erythro and threo forms of 1, 2-bis (4-hydroxy-3-methoxyphenyl)-l, 3-propanediol.
Berndtsson L, et al.
Acta Chemica Scandinavica. Series B, 34, 453-455 (1980)
First enantioselective synthesis of (-)-talaumidin, a neurotrophic diaryltetrahydrofuran-type lignan.
Esumi T, et al.
Tetrahedron Letters, 47(24), 3979-3983 (2006)
(E)-N'-(4-Benzyloxy-3-methoxybenzylidene) benzohydrazide.
He Y-Z and Liu D-Z.
Acta Crystallographica Section E, Structure Reports Online, 61(11), o3855-o3856 (2005)
C A Jackson et al.
Journal of applied microbiology, 122(4), 940-952 (2017-01-17)
The aim of this work was to isolate novel lignin-degrading organisms. Several pure cultures of bacteria that degrade lignin were isolated from bacterial consortia developed from decaying biomass. Among the isolates, Rhizobium sp. strain YS-1r (closest relative of Rhizobium petrolearium
Alla V Lipeeva et al.
European journal of medicinal chemistry, 100, 119-128 (2015-06-17)
A series of 2-(4-R-triazolyl)substituted 3-oxo-2,3-dihydrofurocoumarins have been synthesized by a regioselective cycloaddition of 2-azidooreoselone 1 or 2-azido-9-[(4-methylpiperazin-1-yl)methyl]oreoselone 2 with various alkynes in the presence of Cu(II)/ascorbate in water/methylene chloride reaction medium. The structure of 2-azidooreoselone was established by X-ray structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service