Skip to Content
Merck
All Photos(2)

Key Documents

155411

Sigma-Aldrich

Tetraethyl pyrophosphite

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2POP(OC2H5)2
CAS Number:
Molecular Weight:
258.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.434 (lit.)

bp

79-81 °C/1 mmHg (lit.)

density

1.057 g/mL at 25 °C (lit.)

SMILES string

CCOP(OCC)OP(OCC)OCC

InChI

1S/C8H20O5P2/c1-5-9-14(10-6-2)13-15(11-7-3)12-8-4/h5-8H2,1-4H3

InChI key

RDXABLXNTVBVML-UHFFFAOYSA-N

Application

Tetraethyl pyrophosphite was used in the preparation of diethyl fluorophosphonites.
The product can be used as a reagent for peptide synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service