Skip to Content
Merck
All Photos(2)

Key Documents

151572

Sigma-Aldrich

4-Nitrophenylacetonitrile

98%

Synonym(s):

4-Nitrobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2CN
CAS Number:
Molecular Weight:
162.15
Beilstein:
388442
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

powder

mp

113-115 °C (lit.)

functional group

nitrile
nitro

SMILES string

[O-][N+](=O)c1ccc(CC#N)cc1

InChI

1S/C8H6N2O2/c9-6-5-7-1-3-8(4-2-7)10(11)12/h1-4H,5H2

InChI key

PXNJGLAVKOXITN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenylacetonitrile was used in the preparation of cyanostilbene[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

445. The kinetics of some fast reactions of 4-nitrobenzyl cyanide in solution at ?77?.
Caldin EF and Harbron E.
Journal of the Chemical Society, 2314-2319 (1962)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service