Skip to Content
Merck
All Photos(1)

Key Documents

110930

Sigma-Aldrich

3-Methyl-1,2-butadiene

97%

Synonym(s):

1,1-Dimethylallene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=C=CH2
CAS Number:
Molecular Weight:
68.12
Beilstein:
1697090
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

vapor pressure

6.87 psi ( 20 °C)

Quality Level

Assay

97%

refractive index

n20/D 1.419 (lit.)

bp

40-41 °C (lit.)

mp

−148 °C (lit.)

density

0.694 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)=C=C

InChI

1S/C5H8/c1-4-5(2)3/h1H2,2-3H3

InChI key

PAKGDPSCXSUALC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-1,2-butadiene (1,1-dimethylallene) has been used to study the photo-induced reaction of 1,1-dimethylallene with the cyanoarenes 1,2,4,5-tetracyanobenzene, 1,4-dicyanobenzene and 1,4-dicyanonaphthalene in the presence of methanol as nucleophile.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The electron transfer photochemistry of allenes with cyanoarenes. Photochemical nucleophile?olefin combination, aromatic substitution (photo-NOCAS) and related reactions.
Mangion D, et al.
J. Chem. Soc. Perkin Trans. II, 1, 48-60 (2001)
Shaochen Zhang et al.
Science (New York, N.Y.), 364(6435), 45-51 (2019-04-06)
Accessing enantiomerically enriched amines often demands oxidation-state adjustments, protection and deprotection processes, and purification procedures that increase cost and waste, limiting applicability. When diastereomers can be formed, one isomer is attainable. Here, we show that nitriles, largely viewed as insufficiently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service