Skip to Content
Merck
All Photos(1)

Documents

109363

Sigma-Aldrich

3-Heptanol

99%

Synonym(s):

Butyl ethyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH2CH3
CAS Number:
Molecular Weight:
116.20
Beilstein:
1719067
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020218
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.421 (lit.)

bp

66 °C/20 mmHg (lit.)

density

0.821 g/mL at 20 °C
0.818 g/mL at 25 °C (lit.)

SMILES string

CCCCC(O)CC

InChI

1S/C7H16O/c1-3-5-6-7(8)4-2/h7-8H,3-6H2,1-2H3

InChI key

RZKSECIXORKHQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Heptanol is the main biotransformation product of n-heptane.

Application

3-Heptanol can be used:
  • As a solvent to form microenvironments around single-walled carbon nanotubes.
  • To prepare substituted pyrimidine derivatives as C1 domain-targeted isophthalate analogs to study their binding affinities towards PKCα isoform.
  • As a building block to synthesize 4-(3-adamantan-1-yl-ureido)-butyric acid and cyclohexanecarboxylic acid derivatives as sEH inhibitors.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos A Silvera-Batista et al.
Physical chemistry chemical physics : PCCP, 12(26), 6990-6998 (2010-05-14)
Single-walled carbon nanotubes (SWNTs) are encapsulated with microenvironments of nonpolar solvent, providing a new method to measure the photophysical properties of nanotubes in environments with known properties. Photoluminescence (PL) and absorbance spectra of SWNTs show solvatochromic shifts in 16 nonpolar
Biologically active ester derivatives as potent inhibitors of the soluble epoxide hydrolase
Kim I-H, et al.
Bioorganic & medicinal chemistry letters, 22(18), 5889-5892 (2012)
Solvatochromic shifts of single-walled carbon nanotubes in nonpolar microenvironments
Silvera-Batista CA, et al.
Physical Chemistry Chemical Physics, 12(26), 6990-6998 (2010)
L Perbellini et al.
Archives of toxicology, 58(4), 229-234 (1986-04-01)
Numerous n-heptane metabolites have been identified and quantified by gas chromatography and mass spectrometry in some tissues and in the urine of Sprague Dawley rats exposed for 6 h to 1800 ppm n-heptane. 2-Heptanol and 3-heptanol were the main biotransformation
Scaffold hopping from (5-hydroxymethyl) isophthalates to multisubstituted pyrimidines diminishes binding affinity to the C1 domain of protein kinase C
Provenzani R, et al.
Testing, 13(4), e0195668-e0195668 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service