Skip to Content
Merck
All Photos(3)

Key Documents

109185

Sigma-Aldrich

Saccharin

≥98%

Synonym(s):

2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
Beilstein:
6888
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

mp

226-229 °C (lit.)

solubility

H2O: soluble 1g in 290ml
acetone: soluble 1g in 12ml
alcohol: soluble 1g in 31ml
boiling water: soluble 1g in 25ml
glycerol: soluble 1g in 50ml
alcohol: very slightly soluble
chloroform: slightly soluble
diethyl ether: slightly soluble

SMILES string

O=C1NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)

InChI key

CVHZOJJKTDOEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Saccharin was used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame. It was used in sweet preference test of rats.

Biochem/physiol Actions

Dietery administration of saccharin after N-methyl-N′-nitro-N-nitrosoguanidine promotes tumor development in rats.
A sweet tastant for mammals. A glycerol taste receptor binding site specific for glucose has been proposed in drosophila.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Marie Vinggaard et al.
Toxicological sciences : an official journal of the Society of Toxicology, 85(2), 886-897 (2005-03-25)
Prochloraz is a commonly used fungicide that has shown multiple mechanisms of action in vitro. It antagonizes the androgen and the estrogen receptors, agonizes the Ah receptor, and inhibits aromatase activity. In vivo prochloraz acts antiandrogenically in the Hershberger assay
M Takahashi et al.
Gan, 75(6), 494-501 (1984-06-01)
Sodium chloride, saccharin sodium, phenobarbital sodium and aspirin were tested for tumor-promoting activity in the glandular stomach of rats after initiation with N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) coupled with administration of a high salt diet. Male outbred Wistar rats were given MNNG in
Simultaneous determination of artificial sweeteners in possible counterfeited wines, using high performance liquid chromatography with DAD detection.
Geana EI, et al.
Analele Universitatii Ovidius din Constanta-Seria Chimie, 23(1), 77-81 (2012)
Rino Ragno et al.
Journal of medicinal chemistry, 50(6), 1241-1253 (2007-02-28)
The screening of the inhibition capabilities of dye-like small molecules from a focused library against both human PRMT1 and Aspergillus nidulans RmtA is reported as well as molecular modeling studies (homology modeling, molecular docking, and 3-D QSAR) of the catalytic
Hugh P Morgan et al.
The Biochemical journal, 448(1), 67-72 (2012-08-22)
PYK (pyruvate kinase) plays a central role in the metabolism of many organisms and cell types, but the elucidation of the details of its function in a systems biology context has been hampered by the lack of specific high-affinity small-molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service