Skip to Content
Merck
All Photos(2)

Key Documents

101079

Sigma-Aldrich

3-Bromophenol

98%

Synonym(s):

m-Bromophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OH
CAS Number:
Molecular Weight:
173.01
Beilstein:
1853950
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

236 °C (lit.)

mp

28-32 °C (lit.)

SMILES string

Oc1cccc(Br)c1

InChI

1S/C6H5BrO/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

MNOJRWOWILAHAV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

3-Bromophenol is used for suzuki-miyaura coupling reaction and in the synthesis of pentacyclic building block benzodibenzofuranquinone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium loaded on ZnO nanoparticles: Synthesis, characterization and application as heterogeneous catalyst for Suzuki--Miyaura cross-coupling reactions under ambient and ligand-free conditions
Digambar B B et al.
Materials Chemistry and Physics, 243, 122561-122561 (2020)
Synthesis of a versatile pentacyclic building block for organic electronics
Brenden M et al.
Journal of Polymer Science Part A: Polymer Chemistry, 55, 2618-2628 (2017)
Kesong Tian et al.
Nature communications, 11(1), 3884-3884 (2020-08-06)
Integrating nitrogen species into sp2-hybridized carbon materials has proved an efficient means to improve their electrochemical performance. Nevertheless, an inevitable mixture of nitrogen species in carbon materials, due to the uncontrolled conversion among different nitrogen configurations involved in synthesizing nitrogen-doped
G F Rush et al.
Toxicology, 30(3), 259-272 (1984-04-02)
Bromobenzene, at doses greater than 5.7 mmol/kg, produced renal proximal tubular necrosis and renal functional changes in mice. p-Bromophenol and o-bromophenol were the major urinary phenolic bromobenzene metabolites although m-bromophenol and 4-bromocatechol were also excreted in detectable quantities. With the
D Mahadevan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(2), 575-581 (2010-12-28)
The FT-IR and FT-Raman spectra of 3-Bromo phenol (3-BP) molecule have been recorded using Bruker IFS 66V spectrometer in the range of 4000-100 cm(-1). The molecular geometry and vibrational frequencies in the ground state are calculated by using the ab

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service