Merck
  • Space constraints govern fate of hematopoietic stem and progenitor cells in vitro.
Space constraints govern fate of hematopoietic stem and progenitor cells in vitro.
Biomaterials (2015-04-22)
Eike Müller, Tatyana Grinenko, Tilo Pompe, Claudia Waskow, Carsten Werner
ABSTRACT
Deciphering exogenous cues that determine stem cell fate decisions is a persisting challenge of cell biology and bioengineering. In an effort to unravel the role of spatial constraints in the cell-instructive characteristics of bone marrow microenvironments, murine hematopoietic stem and progenitor cells (HSPC) were exposed to fibronectin-coated microcavities in vitro. Microcavity sizes were chosen to allow for the inclusion of either individual or multiple cells. Repopulation experiments using lethally irradiated mice showed that the maintenance of functional HSPC in culture critically depends on cavity dimensions. Short-term repopulating hematopoietic stem cells (ST-HSC) were found to be best supported within single-cell sized compartments while long-term repopulating HSC (LT-HSC) were maintained within both cavity sizes. In sum, the reported data reveal spatial restriction to be a simple but powerful means for directing HSPC fate ex vivo.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Chlorodimethylsilane, 98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Maleic anhydride, 99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2-Ethyl-2-phenylmalonamide monohydrate, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis