Merck
  • Combinatorialization of fungal polyketide synthase-peptide synthetase hybrid proteins.
Combinatorialization of fungal polyketide synthase-peptide synthetase hybrid proteins.
Journal of the American Chemical Society (2014-12-02)
Thomas B Kakule, Zhenjian Lin, Eric W Schmidt
ABSTRACT
The programming of the fungal polyketide synthase (PKS) is quite complex, with a simple domain architecture leading to elaborate products. An additional level of complexity has been found within PKS-based pathways where the PKS is fused to a single module nonribosomal peptide synthetase (NRPS) to synthesize polyketides conjugated to amino acids. Here, we sought to understand the communication between these modules that enable correct formation of polyketide-peptide hybrid products. To do so, we fused together the genes that are responsible for forming five highly chemically diverse fungal natural products in a total of 57 different combinations, comprising 34 distinct module swaps. Gene fusions were formed with the idea of testing the connection and compatibility of the PKS and NRPS modules mediated by the acyl carrier protein (ACP), condensation (C) and ketoreductase (KR) domains. The resulting recombinant gene fusions were analyzed in a high-yielding expression platform to avail six new compounds, including the first successful fusion between a PKS and NRPS that make highly divergent products, and four previously reported molecules. Our results show that C domains are highly selective for a subset of substrates. We discovered that within the highly reducing (hr) PKS class, noncognate ACPs of closely related members complement PKS function. We intercepted a pre-Diels-Alder intermediate in lovastatin synthesis for the first time, shedding light on this canonical fungal biochemical reaction. The results of these experiments provide a set of ground rules for the successful engineering of hr-PKS and PKS-NRPS products in fungi.
Product Description
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥97.0% (GC)
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Supelco
Pyridine, analytical standard
Supelco
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ≥99.5% (GC)
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%