Merck
  • Potential therapeutic effects of odorants through their ectopic receptors in pigmented cells.
Potential therapeutic effects of odorants through their ectopic receptors in pigmented cells.
Drug discovery today (2017-05-24)
Barbara Pavan, Antonio Capuzzo, Alessandro Dalpiaz
ABSTRACT
Several olfactory receptors (ORs) have been characterized outside the olfactory neuroepithelium in neuronal and non-neuronal tissues, where they were implicated in the recognition of diverse chemical signals. ORs have been found to regulate melanogenesis in skin melanocytes, and OR expression has been found in the human brain nigrostriatal dopaminergic neurons, where production of melanin occurs as neuromelanin and can change with age; OR expression is downregulated in Parkinson's disease. Therefore, we propose several odorants as new functional ligands to ORs expressed in non-olfactory pigmented cells as dopaminergic neurons and melanocytes, where, by acting on cAMP-induced melanin production, they could help to counteract melanogenic and neurodegenerative dysfunctions, including melanoma and Parkinson's disease.
Sigma-Aldrich
HDL mimetic peptide 4F trifluoroacetate, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Methyl-5-(2,2,3-trimethyl-3-cyclopentenyl)pentan-2-ol, ≥90% (GC)