Merck
  • Colon-specific mutual amide prodrugs of 4-aminosalicylic acid for their mitigating effect on experimental colitis in rats.
Colon-specific mutual amide prodrugs of 4-aminosalicylic acid for their mitigating effect on experimental colitis in rats.
European journal of medicinal chemistry (2008-05-13)
Suneela S Dhaneshwar, Mukta Chail, Mahavir Patil, Salma Naqvi, Gaurav Vadnerkar
ABSTRACT
Mutual amide prodrugs of 4-aminosalicylic acid with D-phenylalanine and L-tryptophan were synthesized for targeted drug delivery to the inflamed gut tissue in inflammatory bowel disease. Stability studies in aqueous buffers (pH 1.2 and 7.4) showed that the synthesized prodrugs were stable in both the buffers over a period of 10 h. In rat fecal matter the release of 4-aminosalicylic acid from the prodrugs was in the range of 86-91% over a period of 20 h, with half-lives ranging between 343 and 412 min following first order kinetics. Targeting potential of the carrier system and the ameliorating effect of the amide conjugates were evaluated in trinitrobenzenesulfonic acid-induced experimental colitis model in rats. The prodrugs were assessed for their probable damaging effects on pancreas and liver with the help of histopathological analysis and for their ulcerogenic potential by Rainsford's cold stress method. They were found to have improved safety profile than sulfasalazine, oral 4- and 5-aminosalicylic acid with similar pharmacological spectrum and advantages of sulfasalazine.
Product Description
Supelco
Sulfapyridine, VETRANAL®, analytical standard
Supelco
Sulfapyridine, ≥99.0%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)