Skip to Content
Merck
All Photos(4)

Key Documents

P1751

Sigma-Aldrich

D-Phenylalanine

≥98% (HPLC)

Synonym(s):

(R)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
2804068
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

D-Phenylalanine, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

273-276 °C

application(s)

cell analysis

SMILES string

N[C@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m1/s1

InChI key

COLNVLDHVKWLRT-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Paper Title: </strong> Supramolecular nanofibers co-loaded with dabrafenib and doxorubicin for targeted and synergistic therapy of differentiated thyroid carcinoma. D-Phenylalanine are used to improve the stability of peptides <em>in vivo</em> (Chen P et al., 2023).</li>
<li><strong>Paper Title: </strong> Mechanistic aspects of the transamination reactions catalyzed by D-amino acid transaminase from Haliscomenobacter hydrossis. This study focuses on the enzymatic roles of D-Phenylalanine in specific bacterial transamination processes, applicable in biocatalysis and synthetic biology (Bakunova AK et al., 2023).</li>
</ul>

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jincheng Ni et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(2) (2020-12-30)
Spin angular momentum of light is vital to investigate enantiomers characterized by circular dichroism (CD), widely adopted in biology, chemistry, and material science. However, to discriminate chiral materials with multiscale features, CD spectroscopy normally requires wavelength-swept laser sources as well
Andrew T Schuster et al.
Gastroenterology, 148(7), 1405-1416 (2015-02-24)
Defects in colonic epithelial barrier defenses are associated with ulcerative colitis (UC). The proteins that regulate bacterial clearance in the colonic epithelium have not been completely identified. The Drosophila chromosome-associated protein D3 (dCAP-D3) regulates responses to bacterial infection. We examined
Teresa Blazek et al.
Biosensors & bioelectronics, 163, 112260-112260 (2020-06-23)
The case study of four FAD-dependent oxidase enzymes is presented in the context of the often-claimed direct electron transfer (DET) to glucose oxidase at carbon nanotubes (CNT). The selected enzymes included d-amino acid (AAOx), alcohol (AOx), pyranose (PyOx), and choline
Suneela S Dhaneshwar et al.
European journal of medicinal chemistry, 44(1), 131-142 (2008-05-13)
Mutual amide prodrugs of 4-aminosalicylic acid with D-phenylalanine and L-tryptophan were synthesized for targeted drug delivery to the inflamed gut tissue in inflammatory bowel disease. Stability studies in aqueous buffers (pH 1.2 and 7.4) showed that the synthesized prodrugs were
Ruth Milkereit et al.
Nature communications, 6, 7250-7250 (2015-05-23)
Mammalian target of rapamycin 1 (mTORC1), a master regulator of cellular growth, is activated downstream of growth factors, energy signalling and intracellular essential amino acids (EAAs) such as Leu. mTORC1 activation occurs at the lysosomal membrane, and involves V-ATPase stimulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service