Merck
  • Transient inhibition of rDNA transcription in donor cells improves ribosome biogenesis and preimplantation development of embryos derived from somatic cell nuclear transfer.
Transient inhibition of rDNA transcription in donor cells improves ribosome biogenesis and preimplantation development of embryos derived from somatic cell nuclear transfer.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-04-24)
Chen Liao, Nan Pang, Zhaojun Liu, Lei Lei
ABSTRACT
Ribosomal DNA (rDNA) transcription is a limiting step in ribosome biogenesis, crucial for protein synthesis and cell growth-especially at the early stages of embryonic development-and is regulated in a mammalian target of rapamycin (mTOR)-dependent manner. Our previous report demonstrated that treatment with mTOR inhibitors during artificial embryonic activation improved the development of embryos derived from somatic cell nuclear transfer (SCNT). We hypothesize that inhibition of ribosome biogenesis in somatic cells facilitates reactivation of embryonic nucleolar establishment and ribosome biogenesis in SCNT embryos. Herein, we show that mTOR inhibitors suppressed ribosome biogenesis in somatic cells, and more importantly, improved development potential of SCNT embryos (blastocyst rate, 34% vs 24%). SCNT embryos derived from drug-treated somatic cells exhibited higher levels of 47S, 18S, and 5S rRNAs, upstream binding factor (UBF) mRNA, ribosomal protein S6; they also improved the rebuilding of the nucleolar ultrastructure. In addition, treatment of donor cells with the RNA polymerase I (Pol I) inhibitor cx5461 caused similar effects on SCNT embryos. These results indicated that transient inhibition of rDNA transcription in donor cells facilitated the establishment of functional nucleoli and improved preimplantation development of SCNT embryos.
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, ≥98% (HPLC), powder
Sigma-Aldrich
5-Ethynyl uridine, ≥95%
Supelco
Stainless Steel HPLC Fittings, ferrule, 1/8 in. tubing
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
PP242 hydrate, ≥98% (HPLC), powder