Skip to Content
Merck
All Photos(1)

Key Documents

C6762

Sigma-Aldrich

Cytochalasin B from Drechslera dematioidea

≥98% (HPLC), powder

Synonym(s):

Phomin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37NO5
CAS Number:
Molecular Weight:
479.61
Beilstein:
1096207
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 10 mg/mL

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes

storage temp.

−20°C

SMILES string

C[C@@H]1CCC[C@@H](O)\C=C\C(=O)O[C@]23[C@@H](\C=C\C1)[C@H](O)C(=C)[C@@H](C)[C@H]2[C@H](Cc4ccccc4)NC3=O

InChI

1S/C29H37NO5/c1-18-9-7-13-22(31)15-16-25(32)35-29-23(14-8-10-18)27(33)20(3)19(2)26(29)24(30-28(29)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14-16,18-19,22-24,26-27,31,33H,3,7,9-10,13,17H2,1-2H3,(H,30,34)/b14-8+,16-15+/t18-,19-,22-,23+,24+,26+,27-,29-/m1/s1

InChI key

GBOGMAARMMDZGR-TYHYBEHESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cytochalasin B is a cell-permeable fungal toxin / mycotoxin that binds to the ′barbed′ end of actin / actin filaments. This binding leads to:
  • Disruption of actin filaments and of interaction of actin filaments in solution
  • Inhibition of actin polymerization
  • Inhibition of subunit association and dissociation
Cytochalasin B is widely used in studies of glucose transporters (GLUT). Cytochalasin B is also used as an integral part of various in vitro micronucleus assay protocols.

Biochem/physiol Actions

Cell permeable fungal toxin that disrupts contractile microfilaments by inhibiting actin polymerization. This, in turn, induces DNA fragmentation, inhibits cell division, and disrupts many cell processes. Inhibits glucose transport.
One of a group of fungal metabolites that interfere with a wide variety of cellular movements. Useful tool for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. Probe for the two hexose-transport systems in rat L6 myoblasts.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kinga Brzozowska et al.
International journal of radiation biology, 85(10), 891-899 (2009-07-30)
It has been reported that the level of cytogenetic damage in human peripheral blood lymphocytes (PBL) is higher following irradiation at 37 degrees C than at 0-4 degrees C. The mechanisms of this cytogenetic temperature effect are not fully known.
Christian Johannes et al.
Mutation research, 701(1), 80-85 (2010-05-18)
There are conflicting data regarding the effect of culturing time of human peripheral blood lymphocytes on the yield of chromosomal aberrations induced by sparsely ionising radiation in the G0 phase of the cell cycle. While some authors find that the
N V Ballal et al.
International endodontic journal, 52(8), 1228-1234 (2019-03-09)
To test whether the incorporation of a chelation powder, etidronate, marketed for root canal irrigation (Dual Rinse HEDP) into a sodium hypochlorite (NaOCl) solution induced additional cytotoxic and genotoxic effects not observed with NaOCl alone. Fresh and 24-h-old mixtures of
Matthew Wood et al.
Journal of cell science, 133(20) (2020-09-30)
TAR DNA-binding protein 43 (TDP-43; also known as TARDBP) is an RNA-binding protein whose aggregation is a hallmark of the neurodegenerative disorders amyotrophic lateral sclerosis and frontotemporal dementia. TDP-43 loss increases DNA damage and compromises cell viability, but the actual
Andrea Adamo et al.
Analytical chemistry, 84(15), 6438-6443 (2012-07-04)
Mechanical properties of cells have been shown to have a significant role in disease, as in many instances cell stiffness changes when a cell is no longer healthy. We present a high-throughput microfluidics-based approach that exploits the connection between travel

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service