
3-Formylbenzonitrile

Synonym(s):
3-Cyanobenzaldehyde
Linear Formula:
NCC6H4CHO
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1