Key Documents
K5154-U
Omnifit® EZ Benchmark Replacement Glass Columns
I.D. × L 6.6 mm × 400 mm
About This Item
UNSPSC Code:
23151816
NACRES:
SB.52
Pricing and availability is not currently available.
manufacturer/tradename
Omnifit 006ERG-06-40
Quality Level
technique(s)
LPLC: suitable
I.D. × L
6.6 mm × 400 mm
General description
Replacement glass columns are available in a wide range of bore sizes and lengths. This not only enables you to replace a column should breakage occur, but allows you to obtain several different length columns to use with endpieces you already have for that bore size. The modular nature of our column ranges means that for the ultimate flexibility you can mix and match column sizes with different endpiece types to your specific requirements.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)