Skip to Content
Merck
All Photos(1)

Key Documents

82033-U

Hamilton® PRP-1 (10 µm) HPLC Columns

L × I.D. 20 mm × 2.1 mm, stainless steel hardware

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Hamilton PRP-1 HPLC Guard Cartridge, Kit, 10 μm particle size, L × I.D. 20 mm × 2.1 mm, stainless steel hardware

material

stainless steel hardware

Agency

suitable for USP L21

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

20 mm × 2.1 mm

matrix

polymeric particle platform
polymeric

matrix active group

polymer

particle size

10 μm

pore size

100 Å

operating pH

1-14

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Kit includes 1 holder and 2 cartridges

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service