Skip to Content
Merck
All Photos(1)

Key Documents

47863

Supelco

L-Ascorbic acid

analytical standard

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E300

grade

analytical standard

form

solid

CofA

current certificate can be downloaded

feature

standard type water soluble vitamin

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid plays the role of an antioxidant in plants in regulating the reactive oxygen species mechanism to form reduced peroxides. It is present abundantly as a non-enzymatic antioxidant in higher plants. It can be synthesized in a variety of plants and in all known mammals, except primates and guinea pigs. It can commonly be used in the prevention of scurvy disease.

Application

L-Ascorbic acid may be used as an analytical reference standard for the quantification of the analyte in fruits and vegetable juice samples using flow injection system with immobilised ascorbate oxidase enzyme.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of L-ascorbic acid in fruit and vegetable juices by flow injection with immobilised ascorbate oxidase
Greenway MG and Ongomo P
Analyst, 115(10), 1297-1299 (1990)
Greenberg.MD
Metabolic Pathways: Energetics, Tricarboxylic Acid Cycle, and Carbohydrates, (3), 2014-2014 null
Ascorbic acid deficiency leads to increased grain chalkiness in transgenic rice for suppressed of L-GalLDH
Yu L, et al.
Journal of Plant Physiology, 211, 13-26 (2017)
Takashi Yoshida et al.
Nature chemical biology, 2(11), 596-607 (2006-09-26)
Transient receptor potential (TRP) proteins form plasma-membrane cation channels that act as sensors for diverse cellular stimuli. Here, we report a novel activation mechanism mediated by cysteine S-nitrosylation in TRP channels. Recombinant TRPC1, TRPC4, TRPC5, TRPV1, TRPV3 and TRPV4 of
Marc P Bradshaw et al.
Critical reviews in food science and nutrition, 51(6), 479-498 (2011-09-21)
Extensive reviews of research are available on the use of ascorbic acid, and its consequent degradation pathways, in physiological conditions or food matrices. However, very little information can be found for wine-related systems. This review highlights the relevant chemistry and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service