Key Documents
21992
Supelco
Static Dilution Bottle w/ Mininert® Valve
About This Item
UNSPSC Code:
41104008
packaging
pkg of 1 bottle
technique(s)
active air sampling: suitable
flask volume
2 L
application(s)
air monitoring
environmental
industrial hygiene
General description
  • Static dilution bottle w/ Mininert® valve features a two liter round bottomed flask with a threaded neck.
  • Use the static dilution bottle to prepare gaseous volatile organic standards, using a technique developed by the US EPA for air analyses.
Features and Benefits
MinInert valve included
Legal Information
Mininert is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)