Z326380
BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
volume 5 mL, accuracy: 0.04 mL
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3 flask
wide-neck
Joint
joint ST/NS neck10/19
description
calibrated to contain (TC, IN)
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37401
volume measuring range
5 mL, accuracy: 0.04 mL
capacity
5 mL
General description
The BLAUBRAND® volumetric flask combines precision and reliability, featuring a square grip PP stopper for easy handling. Crafted from high-quality Boro 3.3 glass and calibrated to contain (TC, In), each flask is marked with high-contrast white enamel for clear readability. Supplied with a batch number and certificate, these flasks ensure quality and traceability, making them ideal for laboratory use.
Other Notes
BR37401 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)