Skip to Content
Merck
All Photos(3)

Key Documents

WH0007976M9

Sigma-Aldrich

Monoclonal Anti-FZD3 antibody produced in mouse

clone 2H5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Fz3, Anti-frizzled homolog 3 (Drosophila), Anti-hFz3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD3(7976)

General description

Frizzled class receptor 3 (FZD3) is a receptor with seven-transmembrane domains and it is expressed in human melanocytes. The gene encoding it is localized on human chromosome 8p21.1.

Immunogen

FZD3 (NP_059108, 55 a.a. ~ 157 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QQTAALAMEPFHPMVNLDCSRDFRPFLCALYAPICMEYGRVTLPCRRLCQRAYSECSKLMEMFGVPWPEDMECSRFPDCDEPYPRLVDLNLAGEPTEGAPVAV

Biochem/physiol Actions

Frizzled class receptor 3 (FZD3) modulates the growth of longitudinal axon tracts in the central nervous system. It mediates the dynamics of axon within the enteric, sympathetic and peripheral nervous systems. FZD3 regulates planar cell polarity. Abnormal FZD3 gene methylation causes chromatin structure modifications, associated with congenital hydrocephalus. Mutation in FZD3 gene is associated with Hirschsprung disease, a birth defect lacking the intrinsic ganglion cells of the lower intestine.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Celsr3 and Fzd3 in axon guidance.
The International Journal of Biochemistry & Cell Biology (2015)
Genotype-phenotype association studies of chromosome 8p inverted duplication deletion syndrome.
Fisch GS
Behavior Genetics (2011)
The roles of Frizzled-3 and Wnt3a on melanocyte development: in vitro studies on neural crest cells and melanocyte precursor cell lines.
Chang CH
The Journal of Dermatology (2014)
Impaired methylation modifications of FZD3 alter chromatin accessibility and are involved in congenital hydrocephalus pathogenesis.
Wang L
Brain Research (2014)
Hyunchul Ryu et al.
Nature communications, 12(1), 612-612 (2021-01-29)
The motile cilia of ependymal cells coordinate their beats to facilitate a forceful and directed flow of cerebrospinal fluid (CSF). Each cilium originates from a basal body with a basal foot protruding from one side. A uniform alignment of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service